missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (AISB12), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43435PCP
This item is not returnable.
View return policy
Description
CD20 Monoclonal specifically detects CD20 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD20 | |
| Monoclonal | |
| PerCP | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG2a |
| Flow Cytometry | |
| AISB12 | |
| Flow Cytometry | |
| MS4A1 | |
| The immunogen for this antibody was CD20. | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction