Learn More
CD20 Antibody (B-H20), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-14594AF594
Product Code. 18558494
  • ONLINE EXCLUSIVE PRICE
    € 493.00
    € 466.20 / 0.10mL
    Save € 26.80
    5% Off
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD20
Monoclonal
Alexa Fluor 594
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Chronic Lymphoid Leukaemia cells (CLL)
0.1 mL
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
931
Store at 4°C in the dark.
IgG2a κ
Flow Cytometry
B-H20
50mM Sodium Borate
Mouse
Affinity purified