missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (B-Ly1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28384
This item is not returnable.
View return policy
Description
CD20 Monoclonal specifically detects CD20 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD20 | |
| Monoclonal | |
| APC | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human CD20 | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunoprecipitation, In vitro Assay | |
| B-Ly1 | |
| Flow Cytometry 10 uL/10^6 cells | |
| MS4A1 | |
| The immunogen is a peptide made to the amino acid region CD20 | |
| 1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931 | |
| Human, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction