missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (B9E9), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34559PECY55
This item is not returnable.
View return policy
Description
CD20 Monoclonal specifically detects CD20 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD20 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| MS4A1 | |
| Lymphoblastoid cell line Daudi. | |
| 0.1 mL | |
| Primary | |
| Recognizes a protein of 33-37kDa, identified as CD20 (Workshop V; Code CD20.12). B9E9 recognizes extracellular domain of CD20. The epitope is similar to or identical to that recognized by other CD20 antibodies including Leu-16 and B1. This monoclonal antibody can be used for immunophenotyping of leukemia and malignant cells, B lymphocyte detection in peripheral blood, B cell localization in tissues and B lymphocyte purification by immunosorbent methods. CD20 is a non-Ig differentiation antigen of B-cells and its expression is restricted to normal and neoplastic B-cells, being absent from all other leukocytes and tissues. CD20 is expressed by pre B-cells and persists during all stages of B-cell maturation but is lost upon terminal differentiation into plasma cells. Protein passes through the membrane 4 times with both ends in cytoplasm and exposes one short and one longer loop to the external environment. CD20 is not glycosylated in resting B cells and its cytoplasmic domains are differentially phosphorylated upon activation. It acts as a calcium channel involved in B-cell activation and cell cycle progression. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| B9E9 | |
| Flow Cytometry | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| Mouse | |
| Protein A or G purified | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931.0 | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction