missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD20 Antibody (divozilimab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27978-1mg
This item is not returnable.
View return policy
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD20 | |
| Monoclonal | |
| Unconjugated | |
| P11836 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| divozilimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| CD20 | |
| 1 mg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction