missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (OTI4B4), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70362JF646
This item is not returnable.
View return policy
Description
CD20 Monoclonal specifically detects CD20 in Human, Mouse, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CD20 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MS4A1 | |
| Full length recombinant protein of human CD20 (NP_068769) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Canine, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4B4 | |
| Western Blot, Immunohistochemistry | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| Mouse | |
| Affinity Purified | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction