missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (rMS4A1/8044) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24142JF669
This item is not returnable.
View return policy
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD20 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| Recombinant fragment (around aa200-297) of human CD20 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rMS4A1/8044 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction