Learn More
CD20 Mouse anti-Human, Brilliant Violet 605, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747736
This item is not returnable.
View return policy
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD20 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| CD20 | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 2H7 | |
| Brilliant Violet 605 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG2b κ |