Learn More
CD20 Mouse anti-Human, Brilliant Violet 605, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 747736
Product Code. 17289454
  • ONLINE EXCLUSIVE PRICE
    € 580.00
    € 577.53 / 50µg
    Save € 2.47
    0.43% Off
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD20
Monoclonal
0.2 mg/mL
Mouse
RUO
CD20
Refrigerate (2 to 8°C)
Flow Cytometry
2H7
Brilliant Violet 605
50 μg
Primary
Human
IgG2b κ