Learn More
CD20 Mouse, APC, Clone: 2H7, enQuireBio™
Mouse Monoclonal Antibody
€ 275.10 - € 2268.00
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 2H7 |
| Concentration | 5μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15921828
|
EnquireBio
QAB31-APC-25Tests |
25 Tests |
€ 275.10
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15911828
|
EnquireBio
QAB31-APC-100Tests |
100 Tests |
€ 538.65
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15931828
|
EnquireBio
QAB31-APC-500Tests |
500 Tests |
€ 2268.00
500 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD20 | |
| 5μL/Test | |
| Monoclonal | |
| Mouse | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 931 | |
| IgG2b κ | |
| Protein A/G Purified |
| 2H7 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| NP_690605 | |
| MS4A1 | |
| Primary | |
| Store at 2-4°C. |