missing translation for 'onlineSavingsMsg'
Learn More

CD20 / MS4A1 Mouse, Clone: rIGEL/773, enQuireBio™

Mouse Monoclonal Antibody

€ 199.00 - € 478.00

Specifications

Antigen CD20 / MS4A1
Clone rIGEL/773
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15991188
View Documents
EnquireBio
931-MSM9-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15901198
View Documents
EnquireBio
931-MSM9-P1
100 μg
ONLINE EXCLUSIVE PRICE
€ 554.40
€ 478.00 / 100µg
Save € 76.40
13% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD20 / MS4A1
200μg/mL
Monoclonal
Mouse
10mM PBS with or without 0.05% BSA and 0.05% azide.
931
Recombinant full-length human MS4A1 protein
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
33-37kDa
rIGEL/773
Immunohistochemistry
Unconjugated
RUO
NP_690605
MS4A1
IgG2a κ
Protein A/G Purified
Predominantly cell surface with some cytoplasmic
Daudi, Raji, and U266, and human lymphocytes. Lymph nodes and tonsils.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.