missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200/OX2 Antibody (333), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89696MFV450
This item is not returnable.
View return policy
Description
CD200/OX2 Monoclonal antibody specifically detects CD200/OX2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD200/OX2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 333 | |
| Immunohistochemistry-Paraffin | |
| antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CD200/OX2 (Accession#: NP_005935.4; Met1-Gly232). | |
| 0.1 mL | |
| Immunology | |
| 4345 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction