missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200/OX2 Antibody (6E8B11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52438
This item is not returnable.
View return policy
Description
CD200/OX2 Monoclonal specifically detects CD200/OX2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| CD200/OX2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
| Mouse | |
| 31.3 kDa | |
| 0.1 mg | |
| Immunology | |
| 4345 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA | |
| 6E8B11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CD200 | |
| Purified recombinant fragment of human CD200 (AA: Extra(56-257)) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction