missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200R1 Antibody (019), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90481G
This item is not returnable.
View return policy
Description
CD200R1 Monoclonal specifically detects CD200R1 in Mouse samples. It is validated for ELISA.
Specifications
| CD200R1 | |
| Monoclonal | |
| DyLight 488 | |
| CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD200R1 (NP_067300.1; Met 1-Pro 238). | |
| 0.1 mL | |
| Cancer, GPCR, Neuronal Cell Markers, Neuroscience | |
| 131450 | |
| Mouse | |
| Purified |
| ELISA | |
| 19 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human CD200R1 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction