missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200R1 Antibody (OX108), PE/Atto594 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52642PEATT594
This item is not returnable.
View return policy
Description
CD200R1 Monoclonal antibody specifically detects CD200R1 in Human samples. It is validated for Flow Cytometry
Specifications
| CD200R1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor | |
| Recombinant protein - hCD200RCD4d3+4. | |
| 0.1 mL | |
| Cancer, Neuronal Cell Markers, Neuroscience | |
| 131450 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Flow Cytometry | |
| OX108 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction