missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200R1L Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06395JF549
This item is not returnable.
View return policy
Description
CD200R1L Polyclonal antibody specifically detects CD200R1L in Cynomolgus Monkey samples. It is validated for ELISA
Specifications
| CD200R1L | |
| Polyclonal | |
| ELISA | |
| cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa | |
| Produced in rabbits immunized with purified, recombinant Cynomolgus CD200R1L (Met 1-Leu 240). | |
| 0.1 mL | |
| Primary | |
| Cynomolgus Monkey | |
| Purified |
| ELISA | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 344807 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction