missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200R4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06092-100ul
This item is not returnable.
View return policy
Description
CD200R4 Polyclonal antibody specifically detects CD200R4 in Mouse samples. It is validated for ELISA
Specifications
| CD200R4 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| F630107N04Rik, MCD200, MCD200RLa | |
| Produced in rabbits immunized with purified, recombinant Mouse CD200R4 (Accession#: NP_997127.1; Met 1-Thr 241) | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 239849 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction