missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD206 (MMR) Mouse anti-Human, BUV615, Clone: 15-2 (also known as MR15-2), BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751782
This item is not returnable.
View return policy
Description
CD206 (MMR) Monoclonal antibody specifically detects CD206 (MMR) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD206 (MMR) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Hu CD206 (MMR) BUV615 15-2 50 μg | |
| Purified Human MMR | |
| RUO | |
| CD206 (MMR) | |
| Refrigerate (2 to 8°C) | |
| Human |
| Flow Cytometry | |
| 15-2 (also known as MR15-2) | |
| Brilliant Ultraviolet 615 | |
| Mouse | |
| 1 x 50 μg | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction