Learn More
CD206 Mouse anti-Human, FITC, Clone: 15-2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech FITC-65155-100TESTS
Product Code. 17254866
  • € 118.00 / 100 reactions
Description
CD206, also named as MMR, CLEC13D and MRC1, is a type I membrane receptor that mediates the endocytosis of glycoproteins by macrophages. CD206 has been shown to bind high-mannose structures on the surface of potentially pathogenic viruses, bacteria, and fungi so that they can be neutralized by phagocytic engulfment. CD206 is a 170 kDa transmembrane protein which contains 5 domains: an amino-terminal cysteine-rich region, a fibronectin type II repeat, a series of eight tandem lectin-like carbohydrate recognition domains (responsible for the recognition of mannose and fucose), a transmembrane domain, and an intracellular carboxy-terminal tail. It is expressed on most tissue macrophages, in vitro derived dendritic cells, lymphatic and sinusoidal endothelia.
Specifications
Specifications
CD206
Monoclonal
4 μL/Test
Mouse
Primary
Human, Mouse
IgG1 κ
Flow Cytometry
15-2
FITC
100 Tests
4360