missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD206 Rat anti-Mouse, Alexa Fluor 647, Clone: MR5D3, BD Pharmingen™
Rat Monoclonal Antibody
Brand: BD Biosciences 565250
This item is not returnable.
View return policy
Description
CD206 Monoclonal antibody specifically detects CD206 in Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
| CD206 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 25 μg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
| MR5D3 | |
| Alexa Fluor 647 | |
| MMR; MR; Macrophage mannose receptor 1; Mrc1; Mannose receptor, C type 1 | |
| Recombinant Mouse CD206 Carbohydrate recognition domains 4-7/Fc Fusion Protein | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. | |
| IgG2a, CDF IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction