missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Antibody (21A/5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50178PE
This item is not returnable.
View return policy
Description
CD21 Monoclonal specifically detects CD21 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD21 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| CR2 | |
| CD21 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Dendritic Cell Markers, Immunology | |
| 1380 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| 21A/5 | |
| Flow Cytometry | |
| C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction