missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Antibody (544408), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4909W
This item is not returnable.
View return policy
Description
CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| CD21 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 544408 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD21, Ile21-Arg971, Accession # P20023 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Dendritic Cell Markers, Immunology | |
| 1380 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction