missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Antibody (Bu33), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63574AF750
This item is not returnable.
View return policy
Description
CD21 Monoclonal specifically detects CD21 in Human, Equine samples. It is validated for Flow Cytometry.
Specifications
| CD21 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CR2 | |
| Made to Human CD21 | |
| Primary | |
| NB100-63574 recognizes human CD21, a 145kD cell surface glycoprotein expressed by mature B lymphocytes, follicular dendritic cells, subsets of normal thymocytes and subsets of T cells. The CD21 molecule acts as a receptor for CD23, for complement components C3d, C3dg and iC3b and for Epstein Barr Virus. CD21 is part of the signalling complex that is linked to CD19, CD81 and phosphatidylinositol-3-kinase. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| Bu33 | |
| Flow Cytometry | |
| C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
| Mouse | |
| 0.1 mL | |
| 1380 | |
| Human, Equine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction