missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD21 Antibody (CL14552), Novus Biologicals™
Mouse Monoclonal Antibody
€ 387.00
Specifications
| Antigen | CD21 |
|---|---|
| Clone | CL14552 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043660
|
Novus Biologicals™
NBP3-26866-25ul |
25 μL |
€ 387.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043533
|
Novus Biologicals™
NBP3-26866-100ul |
100 μL |
N/A
|
N/A | |||||
Description
CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| CD21 | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Dendritic Cell Markers, Immunology | |
| PBS, pH 7.2, 40% glycerol | |
| 1380 | |
| IgG1 | |
| Protein A purified |
| CL14552 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
| This antibody was developed against a Synthetic Peptide of P20023. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title