missing translation for 'onlineSavingsMsg'
Learn More

CD21/CD35 Rat anti-Mouse, FITC, Clone: 7G6, BD

Rat Monoclonal Antibody

€ 160.66 - € 294.25

Specifications

Antigen CD21/CD35
Clone 7G6
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15841319
View Documents
BD Biosciences
561769
0.1 mg
ONLINE EXCLUSIVE PRICE
€ 161.00
€ 160.66 / Each
Save € 0.34
0.21% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15887408
View Documents
BD Biosciences
553818
0.5 mg
ONLINE EXCLUSIVE PRICE
€ 296.00
€ 294.25 / Each
Save € 1.75
0.59% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 7G6 antibody recognizes an epitope shared by 145-150-kDa and 190-kDa complement receptor proteins, originally designated CR2 (CD21) and CR1 (CD35), respectively. In the mouse, CD21 and CD35 are expressed on the majority of peripheral B lymphocytes, on the majority of resident peritoneal macrophages and mast cells, on peripheral blood granulocytes after treatment with N-formyl-Met-Leu-Phe, and on follicular dendritic cells, but not on thymocytes, T cells, erythrocytes, or platelets. CD21 is a ligand-binding component of the CD19/CD21/CD81 signal-transduction complex associated with the antigen receptor on B lymphocytes. CD21/CD35 also co-localizes with CD19 on the surface of peritoneal mast cells.Cr2null mice display impaired inflammatory and humoral immune responses in vivo. The 7G6 mAb has been reported to inhibit rosette formation by C3d-bearing sheep erythrocytes, to block the complement dependent trapping of immune complexes by follicular dendritic cells, and to down-regulate mouse CD21/CD35 expression upon in vivo application, thus inhibiting primary antibody responses to immunization. Co-stimulation of B-cell differentiation via Sepharose-coupled 7G6 antibody has also been observed. The 7G6 mAb recognizes an epitope on CD35 distinct from the epitope recognized by anti-mouse CD35, clone 8C12 (Cat. No. 558768, for the purified antibody), and it does not block binding of 8C12 mAb to mouse CD35.

Specifications

Specifications

CD21/CD35
0.5mg/mL
Monoclonal
Rat
Mouse
Purified Mouse CR1
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
7G6
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2b κ
Affinity Purified
CR2/CR1
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.