missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD21 Recombinant Rabbit Monoclonal Antibody (257), PE, Invitrogen™
GREENER_CHOICE

Product Code. 30019625
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019625 100 Tests Pack of 100
30019979 25 Tests Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30019625 Supplier Invitrogen™ Supplier No. MA552254

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CD21 (complement receptor 2, CR2, C3D receptor, EBV receptor) binds C3 complement fragments, especially its breakdown fragments, which remain covalently attached to complement activating surfaces or antigen. CD21 has important roles in uptake and retention of immunocomplexes, survival of memory B cells and in development and maintenance of the humoral response to T-dependent antigens. CD21 also serves as a key receptor for Epstein-Barr virus binding and is involved in targeting prions to follicular dendritic cells and expediting neuroinvasion following peripheral exposure to prions. A soluble form of the CD21 (sCD21) is shed from the lymphocyte surface and retains its ability to bind respective ligands. CD21 functions as receptor for C3d, C3dg and iC3b complement components, for EBV and for IFNalpha. CD21 binds to CD23 and associates with CD19, CD81 and Leu13 to form a large signal-transduction complex involved in B cell activation. Genetic variations in the CD21 gene are associated with susceptibility to systemic lupus erythematosus type 9 (SLEB9). Alternatively, spliced transcript variants encoding different isoforms of CD21 have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD21
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 257
Concentration 100 μg/mL
Conjugate PE
Formulation PBS with 0.05% BSA and 0.09% sodium azide
Gene CR2
Gene Accession No. P20023
Gene Alias C3b/C4b receptor; C3-binding protein; C3BR; C3DR; C4BR; CD21; CD35; CD35 antigen; cell surface receptor for C3d; complement C3b/C4b receptor 1 (Knops blood group); complement C3d receptor; Complement C3d receptor (C3DR); complement C3d receptor 2; complement component (3b/4b) receptor 1 (Knops blood group); complement component (3d/Epstein Barr virus) receptor 2; complement component 3b/4b receptor 1 (Knops blood group); complement component 3d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; complement receptor type 1-like protein; complement receptor type 2; Complement Receptor type 2 (CR2); CR; CR1; Cr-1; CR1-L; Cr2; Cr-2; CVID7; EBV receptor; EBV-R; Epstein-Barr virus receptor; EVBR; expressed in B lymphocytes; KN; Knops blood group antigen; LOW QUALITY PROTEIN: complement receptor type 1; SLEB9
Gene Symbols CR2
Host Species Rabbit
Immunogen Recombinant Human CD21 Protein (Ile 21-Arg971).
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1380
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.