missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD22 Mouse anti-Human, Clone: BLCAM/1795, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15056.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against partial recombinant human CD22.
- High specificity
- High reactivity
Specifications
| CD22 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined b | |
| P20273 | |
| CD22 | |
| Recombinant protein corresponding to amino acids 52-178 of human CD22. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| BLCAM/1795 | |
| Mouse monoclonal antibody raised against partial recombinant human CD22. | |
| In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| FLJ22814/MGC130020/SIGLEC-2/SIGLEC2 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 933 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction