missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD22 Mouse anti-Human, Clone: OTI8E6, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807844S
This item is not returnable.
View return policy
Description
CD22 Monoclonal antibody specifically detects CD22 in Human samples. It is validated for Western BlotSpecifications
| CD22 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P20273 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 933 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI8E6 | |
| Unconjugated | |
| CD22 | |
| A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14 | |
| Human recombinant protein fragment corresponding to amino acids 707-847 of human CD22 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction