missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD223 (LAG-3) Monoclonal Antibody (3DS223H), Brilliant Violet™ 605, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
€ 188.00 - € 385.00
Specifications
| Antigen | CD223 (LAG-3) |
|---|---|
| Clone | 3DS223H |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17902021
|
Invitrogen™
406-2239-41 |
25 Tests |
€ 188.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17962111
![]() |
Invitrogen™
406-2239-42 |
100 Tests |
€ 385.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).Specifications
| CD223 (LAG-3) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3 | |
| LAG3 | |
| IgG1 κ | |
| Affinity chromatography | |
| LAG3 |
| 3DS223H | |
| Flow Cytometry | |
| Brilliant Violet 605 | |
| Mouse | |
| Human | |
| P18627 | |
| 3902 | |
| hCD223-FC PFA 4%. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title