Learn More
Invitrogen™ CD226 (DNAM-1) Monoclonal Antibody (DX11)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA528149
Product Code. 16382444
  • € 752.00 / 500µg
Description
This antibody recognizes human CD226, approximately a 65 kDa glycoprotein, also known as DNAM1 (DNAX accessory molecule-1). CD226 is broadly expressed on T-cells, NK cells, platelets, monocytes and a subset of B cells.
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD226 (DNAM-1)
Monoclonal
1 mg/mL
PBS with no preservative
Q15762
Cd226
Human cytotoxic T lymphocyte clone.
500 μg
Primary
Human
Antibody
IgG1
Flow Cytometry, Functional Assay, Immunoprecipitation
DX11
Unconjugated
Cd226
adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1
Mouse
Protein A
RUO
10666
-20°C, Avoid Freeze/Thaw Cycles