missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD226 (DNAM-1) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598421
This item is not returnable.
View return policy
Description
CD226 (DNAM-1) Polyclonal Antibody for Western Blot, IHC (P), ELISA
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
Specifications
| CD226 (DNAM-1) | |
| Polyclonal | |
| Unconjugated | |
| Cd226 | |
| adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10666 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.9 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15762 | |
| Cd226 | |
| Recombinant Human CD226 antigen protein (19-254AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction