missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD226 (DNAM-1) Recombinant Rabbit Monoclonal Antibody (BLR063G)
Rabbit Recombinant Monoclonal Antibody
€ 156.00 - € 474.00
Specifications
| Antigen | CD226 (DNAM-1) |
|---|---|
| Clone | BLR063G |
| Concentration | 1 mg/mL |
| Content And Storage | 4°C |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17755689
|
Invitrogen™
MA544319 |
100 μL |
€ 474.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17765689
|
Invitrogen™
MA544320 |
10 μL |
€ 156.00
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.Specifications
| CD226 (DNAM-1) | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.09% sodium azide; pH 8.2 | |
| Q15762 | |
| 10666 | |
| Recombinant protein representing the extracellular domain of human DNAM-1/CD226 (residues 19-229). | |
| Antibody |
| BLR063G | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Cd226 | |
| adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1 | |
| Cd226 | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title