missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD226 Recombinant Rabbit Monoclonal Antibody (026), APC
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA546800

Product Code. 17830556

  • € 431.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD226 (DNAM-1) (Met1-Pro254) and conjugated with APC under optimum conditions, the unreacted APC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.

DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD226
Recombinant Monoclonal
100 μg/mL
PBS with 0.5% BSA and 0.09% sodium azide
Q8K4F0
Cd226
Recombinant protein encoding Mouse CD226 (Met1-Pro254).
100 Tests
Primary
Mouse
Antibody
IgG
Flow Cytometry
026
APC
Cd226
adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1
Rabbit
Protein A
RUO
225825
4°C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.