Learn More
Invitrogen™ CD226 Recombinant Rabbit Monoclonal Antibody (026), PE

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA546802
Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD226 (DNAM-1) (Met1-Pro254) and conjugated with PE under optimum conditions, the unreacted PE was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
Specifications
| CD226 | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| Q8K4F0 | |
| Cd226 | |
| Recombinant protein encoding Mouse CD226 (Met1-Pro254). | |
| 100 Tests | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 026 | |
| PE | |
| Cd226 | |
| adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 225825 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.