missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ B1 Bradykinin Receptor Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5148223

Product Code. 30019238

  • € 361.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

B1 Bradykinin Receptor Polyclonal Antibody for Western Blot

Bradykinin, a 9 aa peptide, is generated in pathophysiologic conditions such as inflammation, trauma, burns, shock, and allergy. Two types of G-protein coupled receptors have been found which bind bradykinin and mediate responses to these pathophysiologic conditions. The protein encoded by this gene is one of these receptors and is synthesized de novo following tissue injury. Receptor binding leads to an increase in the cytosolic calcium ion concentration, ultimately resulting in chronic and acute inflammatory responses.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

B1 Bradykinin Receptor
Polyclonal
Unconjugated
Bdkrb1
B1 bradykinin receptor; B1BKR; B1R; Bdkrb; BDKRB1; BDKRB2; Bk b1 receptor; BK-1 receptor; BKB1R; BKR; BKR1; BKRB1; BRADYB1; bradykinin B1 receptor; bradykinin receptor 1; bradykinin receptor B1; bradykinin receptor B2; bradykinin receptor, beta 1; KB1; kinin B1; kinin B1 receptor
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
623
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
100 μg/mL
PBS with 0.03% ProClin 300; pH 7
P46663
Bdkrb1
A synthetic peptide corresponding to the N-terminus of the Human BDKRB1.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.