missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (MM0163-3L25), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12151APCCY7
This item is not returnable.
View return policy
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD229/SLAMF3/Lymphocyte Antigen 9 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| LY9 | |
| Human recombinant CD229/SLAMF3/Lymphocyte Antigen 9 (extracellular domain) | |
| 0.1 mL | |
| 4063.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| MM0163-3L25 | |
| Flow Cytometry | |
| CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction