missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (MM0163-3L25), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12151G
This item is not returnable.
View return policy
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| CD229/SLAMF3/Lymphocyte Antigen 9 | |
| Monoclonal | |
| DyLight 488 | |
| Q9HBG7 | |
| LY9 | |
| Human recombinant CD229/SLAMF3/Lymphocyte Antigen 9 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| MM0163-3L25 | |
| Western Blot, Flow Cytometry | |
| CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 4063 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction