missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD23/Fc epsilon RII Antibody (FCER2/8236R), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24059MFV450
This item is not returnable.
View return policy
Description
CD23/Fc epsilon RII Monoclonal antibody specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD23/Fc epsilon RII | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor | |
| Recombinant fragment (around aa 200-300) of human CD23/Fc epsilon RII protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Immunology, Phospho Specific | |
| 2208 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| FCER2/8236R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction