missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD23 Monoclonal Antibody (EBVCS2), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
€ 178.00 - € 392.00
Specifications
| Antigen | CD23 |
|---|---|
| Clone | EBVCS2 |
| Concentration | 0.4 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089956
|
Invitrogen™
H063T02Y04-A |
25 Tests |
€ 178.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089970
|
Invitrogen™
H063T03Y04-A |
100 Tests |
€ 392.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD23 Monoclonal antibody specifically detects CD23 in Human samples. It is validated for Flow CytometrySpecifications
| CD23 | |
| 0.4 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| BLAST-2; CD23; CD23 antigen; CD23A; CLEC4J; C-type lectin domain family 4 member J; C-type lectin domain family 4, member J; Fc epsilon receptor II; FC epsilon RII; Fc fragment of IgE receptor II; Fc fragment of IgE, low affinity II, receptor for (CD23); Fc receptor alpha multiple ligand receptor; Fc receptor, IgE, low affinity II, alpha polypeptide; Fcalpha muR; Fcalpha RII; Fce2; fc-epsilon-RII; FCER2; Fcer2a; FceRII; IGEBF; Immunoglobulin E-binding factor; immunoglobulin epsilon-chain; low affinity immunoglobulin epsilon Fc receptor; Low affinity immunoglobulin epsilon Fc receptor membrane-bound form; Low affinity immunoglobulin epsilon Fc receptor soluble form; low-affinity IgE receptor; Ly-42; lymphocyte IgE receptor | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| EBVCS2 | |
| Flow Cytometry | |
| NovaFluor Yellow 660 | |
| Mouse | |
| Human | |
| P06734 | |
| 2208 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| FCER2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title