Learn More
CD23 Mouse anti-Human, Brilliant Violet 510, Clone: EBVCS-5 (also known as EBVCS 5), BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 743427
Description
The EBVCS-5 antibody specifically binds to human CD23, the low affinity receptor for human IgE (Fc epsilon RII orFcεRII), which is also known as C-type lectin domain family 4 member J (CLEC4J). CD23 is expressed as a ñ45 kDa type II membrane glycoprotein that is present at low density on most normal B lymphocytes and at higher levels on activated B lymphocytes, Epstein-Barr virus (EBV)-transformed lymphoblasts, chronic lymphocytic leukemia (CLL) cells of B-lymphocyte origin, and tonsillar B lymphocytes. CD23 expression is induced on B cells by interleukin-4 (IL-4) and is lost after isotype switching to IgA, IgG, or IgE. The CD23 antigen is not present on immature bone marrow B lymphocytes or on T lymphocytes. CD23 may also be differentially expressed on monocytes, macrophages, eosinophils, platelets, dendritic cells, and Langerhans cells. CD23 can mediate IgE-dependent cytotoxicity and phagocytosis by macrophages and eosinophils. Soluble CD23 (sCD23) can be released by CD23-positive cells as a result of proteolytic cleavage of membrane CD23. Larger fragments of sCD23 (eg, 25-37 kDa) retain their IgE-binding capacity whereas smaller fragments (ie, ≤ 12 kDa) do not. Soluble CD23 may have immunoregulatory effects on the growth and differentiation of B cells and other cell types.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
| CD23 | |
| EBVCS-5 (also known as EBVCS 5) | |
| Brilliant Violet 510 | |
| CD23A, Fc-epsilon-RII, FCER2, FCE2, IGEBF, BLAST-2, CLEC4J, Leu-20 | |
| Epstein-Barr Virus Transformed Cell Line | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.