missing translation for 'onlineSavingsMsg'
Learn More

CD235ab (Glycophorin A/B) Mouse, anti-Human, FITC, Clone: HIR2 (GA-R2), STEMCELL Technologies

Product Code. 17189101
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17189101 25 μg 25µg
17179101 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17189101 Supplier Stemcell Technologies Supplier No. 60111FI.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

  • The HIR2 (GA-R2) antibody recognizes an epitope common to the N-terminal region of human CD235a (glycophorin A) and CD235b (glycophorin B), homologous type I sialoglycoproteins of the erythrocyte (red blood cell) membrane that bear the antigenic determinants for the MN and Ss blood groups
  • CD235ab is expressed on early- and late-stage erythroblasts, mature erythrocytes, and erythroid cell lines such as K562 and HEL, but not on other cell types
  • CD235a is abundantly expressed whereas CD235b is a relatively minor membrane component
  • The proteins are believed to provide a large mucin-like surface to erythrocytes that acts to minimize aggregation in the circulation
  • The HIR2 antibody binds with higher affinity to CD235a than CD235b, and agglutinates untreated erythrocytes

Specifications

Antigen CD235ab (Glycophorin A/B)
Applications Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone HIR2 (GA-R2)
Conjugate FITC
Host Species Mouse
Quantity 25 μg
Primary or Secondary Primary
Target Species Human
Isotype IgG2b κ
Cell Type Red Blood Cells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.