missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD235ab (Glycophorin A/B) Mouse, anti-Human, FITC, Clone: HIR2 (GA-R2), STEMCELL Technologies
Description
- The HIR2 (GA-R2) antibody recognizes an epitope common to the N-terminal region of human CD235a (glycophorin A) and CD235b (glycophorin B), homologous type I sialoglycoproteins of the erythrocyte (red blood cell) membrane that bear the antigenic determinants for the MN and Ss blood groups
- CD235ab is expressed on early- and late-stage erythroblasts, mature erythrocytes, and erythroid cell lines such as K562 and HEL, but not on other cell types
- CD235a is abundantly expressed whereas CD235b is a relatively minor membrane component
- The proteins are believed to provide a large mucin-like surface to erythrocytes that acts to minimize aggregation in the circulation
- The HIR2 antibody binds with higher affinity to CD235a than CD235b, and agglutinates untreated erythrocytes
Specifications
Specifications
| Antigen | CD235ab (Glycophorin A/B) |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | HIR2 (GA-R2) |
| Conjugate | FITC |
| Host Species | Mouse |
| Quantity | 25 μg |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgG2b κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?