Learn More
CD238 Mouse anti-Human, BUV395, Clone: BRIC 203, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742676
Description
The BRIC 203 monoclonal antibody specifically binds to the Kell blood group antigen, Kp(a-b+) but not Kp(a+b-), also known as the Kell blood group metallo-endopeptidase, KELL, ECE3 and CD238. CD238 is an approximately 115-kDa type II transmembrane glycoprotein that is encoded by the KEL gene. CD238 functions as a zinc endopeptidase that cleaves endothelin-3 into an active form which can function as a vasoconstrictor. CD238 is expressed on erythrocytes, human embryonic stem cell-derived endoderm and on cells from various other tissues, including testis, heart, brain and skeletal muscle. Decreased expression of CD238 is associated with the McLeod phenotype as a consequence of losing XK protein expression. Kell antigen expression can play an important role during blood transfusions and also specific autoimmune diseases related to Kell antigen recognition. During development the BRIC 203 monoclonal antibody was found to detect the CD238 antigen by flow cytometric analysis of human erythrocytes. It reportedly works in immunoprecipitation and agglutination assays.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications
| CD238 | |
| BRIC 203 | |
| Brilliant Ultraviolet 395 | |
| KEL, KELL, Kell blood group, metallo-endopeptidase, ECE3 | |
| Human erythrocytes | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.