missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Antibody (ML5), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-77903PCP
This item is not returnable.
View return policy
Description
CD24 Monoclonal antibody specifically detects CD24 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| CD24 | |
| Monoclonal | |
| PerCP | |
| CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
| The immunogen was the full length version of the protein: CD24 (ML5) | |
| 0.1 mL | |
| Cancer, Neuronal Cell Markers, Stem Cell Markers | |
| 100133941 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen) | |
| ML5 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction