missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Mouse anti-Human, BUV395, Clone: ML5, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566221
This item is not returnable.
View return policy
Description
CD24 Monoclonal antibody specifically detects CD24 in Human samples. It is validated for Flow cytometry.
Specifications
| CD24 | |
| Monoclonal | |
| Brilliant Ultraviolet 395 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity and unconjugated antibody and free BD Horizon™ BUV395 were removed. chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions | |
| IgG2a κ |
| Flow Cytometry | |
| ML5 | |
| Vol. Per test: 5μL | |
| Heat Stable Antigen Homologue (HSA); Ba-1; CD24A | |
| 25 Tests | |
| Cell Biology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction