Learn More
CD24 Rat anti-Mouse, PE, Clone: M1/69, BD
Rat Monoclonal Antibody
€ 90.54 - € 140.00
Specifications
| Antigen | CD24 |
|---|---|
| Clone | M1/69 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15860749
|
BD Biosciences
561079 |
25 μg |
€ 90.95 € 90.54 / Each Save € 0.41 0.45% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15857108
|
BD Biosciences
553262 |
0.1 mg |
€ 140.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The M1/69 monoclonal antibody specifically binds to CD24 (Heat-Stable Antigen, HSA or HsAg), a variably glycosylated, glycosyl-phosphatidylinositol-anchored membrane protein expressed on erythrocytes, granulocytes, monocytes, lymphocytes, and neurons. Hematopoietic stem cells of the embryonic yolk sac and fetal liver express CD24. Levels of expression of CD24 vary during differentiation of the T and B cell lineages. Inthe bone marrow, hematopoietic progenitors acquire CD24 expression upon commitment to the B-lymphocyte lineage. Immature B cells in the bone marrow express low CD24 levels whereas peripheral B lymphocytes express intermediate to high levels of CD24. The level of CD24 expression has been reported to rise upon activation of splenic B cells with LPS, but not with CD154 (CD40 Ligand). The majority of thymocytes express high levels of CD24, while most mature thymic and peripheral T lymphocytes do not express CD24. In contrast, TCR-bearing thymocytes which emigrate to the spleen are CD24+. Dendritic cells of the thymus, spleen, liver, and epidermal Langerhans cells have also been reported to express CD24. CD24 is not expressed by NK cells, as determined by staining with J11d mAb (Cat. No. 553146). CD24 is involved in the costimulation of CD4+ T cells by B cells, it is a “co-inducer” of in vitro thymocyte maturation, and it is a ligand of CD62P (P-selectin). While the monoclonal antibodies 30-F1, M1/69, and J11d all react with CD24, they show subtle differences in the level of staining of different lymphocyte populations. When possible, investigators should continue to use the same monoclonal anti-CD24 antibody as used in previous studies.
Specifications
| CD24 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| C57BL/10 Mouse Splenic T Lymphocytes | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| M1/69 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2b κ | |
| Affinity Purified | |
| CD24a; HSA; Heat Stable Antigen; Ly-52; Nectadrin; R13-Ag |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.