missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD248 Monoclonal Antibody (OTI4B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807122
This item is not returnable.
View return policy
Description
CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.Specifications
| CD248 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HCU0 | |
| CD248 | |
| Human recombit protein fragment corresponding to amino acids 20-300 of human CD248 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI4B8 | |
| Unconjugated | |
| CD248 | |
| CD164L1, TEM1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57124 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction