Learn More
CD248 Mouse anti-Human, Clone: OTI4B8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807122
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.Specifications
| CD248 | |
| Monoclonal | |
| Unconjugated | |
| CD248 | |
| CD164L1, TEM1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57124 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4B8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9HCU0 | |
| CD248 | |
| Human recombit protein fragment corresponding to amino acids 20-300 of human CD248 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.