missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25/IL-2R alpha Antibody (280414), Alexa Fluor 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals MAB24383AF750
This item is not returnable.
View return policy
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD25/IL-2R alpha | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IL2RA | |
| Partial recombinant mouse CD25/IL2Ra protein (amino acids 22-236) [UniProt P01590]. | |
| Primary | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| 280414 | |
| Flow Cytometry | |
| CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| Rat | |
| 0.1 mL | |
| 3559 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction