Learn More
CD25/IL-2R alpha Antibody (745520), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB51561PECY7
Product Code. 30028124
  • € 618.00 / 0.10mL
Description
CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD25/IL-2R alpha
Monoclonal
PE-Cyanine7
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Rat
Purified
Flow Cytometry
745520
Flow Cytometry
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Chinese hamster ovary cell line CHO-derived recombinant rat CD25/IL-2 R alpha , Glu22-Gln235, Accession # P26897
0.1 mL
Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction
3559
Store at 4C in the dark. Do not freeze.