missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25/IL-2R alpha Antibody (PC61.5.3), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB9164MFV610
This item is not returnable.
View return policy
Description
CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CD25/IL-2R alpha | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| PC61.5.3 | |
| Flow Cytometry, CyTOF-ready | |
| CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| B6.1 mouse cytotoxic T cell line, Accession # P01590 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
| 3559 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction